2-Imidazolidinone ≥96%

Supplier: BeanTown Chemical

Synonyms: 2-Imidazolidone

135365-1KG 135365-500G
BT135365-1KGEA 169 USD
BT135365-1KG BT135365-500G
2-Imidazolidinone ≥96%
2-Imidazolidinone

CAS: 120-93-4; EC No: 204-436-4; MDL No: MFCD00005257; RTECS: NJ0570000 Powder; Molecular Formula: C3H6N2O; MW: 86.09 Melting Point: 129-132°


  • RTECS: NJ0570000
  • Powder

Formula: C₃H₆N₂O
MW: 86.0925 g/mol
Melting Pt: 129…132 °C
MDL Number: MFCD00005257
CAS Number: 120-93-4
EINECS: 204-436-4

Order Now

Specification Test Results

Assay 96%

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR