4-Chloro-N-methylpicolinamide ≥97%
Supplier: BeanTown Chemical
135330-1G
135330-250MG
BT135330-1GEA
253
USD
BT135330-1G
BT135330-250MG
4-Chloro-N-methylpicolinamide ≥97%
4-Chloro-N-methylpicolinamide
CAS: 220000-87-3; MDL No: MFCD02185921
Solid; Molecular Formula: C7H7ClN2O ; MW: 170.60
Melting Point: 34-43°
- Solid
Formula:
C₇H₇ClN₂O Melting Pt: 34…43 °C |
MDL Number:
MFCD02185921 CAS Number: 220000-87-3 |
Specification Test Results
Assay | 97% |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...