Calcium trifluoromethanesulfonate ≥99%
Supplier: BeanTown Chemical
Synonyms:
Calcium trifluoromethanesulphonate, Calcium triflate
135325-10G
135325-50G
BT135325-10GEA
83.98
USD
BT135325-10G
BT135325-50G
Calcium trifluoromethanesulfonate ≥99%
Calcium trifluoromethanesulphonate
CAS: 55120-75-7; EC No: 232-188-7; MDL No: MFCD00143911
Powder; Linear Formula: (CF3SO3)2Ca; MW: 338.21
Melting Point: 350°
- Powder
Formula:
C₂CaF₆O₆S₂ MW: 338.22 g/mol Melting Pt: 350 °C |
MDL Number:
MFCD00143911 CAS Number: 55120-75-7 EINECS: 232-188-7 |
Specification Test Results
Assay | 99% |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...