Allylpalladium(II) chloride dimer

Supplier: BeanTown Chemical

Synonyms: Di-mu-chlorobis[(eta-allyl)palladium(II)], Allylpalladium(II) chloride, Allylpalladium chloride dimer

130490-1G 130490-250MG 130490-5G
BT130490-1GEA 133 USD
BT130490-1G BT130490-250MG BT130490-5G
Allylpalladium(II) chloride dimer
Allylpalladium(II) chloride dimer

CAS: 12012-95-2; EC No: 234-579-8; MDL No: MFCD00044874 Powder; Molecular Formula: C6H10Cl2Pd2; MW: 365.89 Air Sensitive, Moisture Sensitive


  • Air Sensitive, Moisture Sensitive
  • Powder

Formula: C₆H₁₀Cl₂Pd₂
MW: 365.89 g/mol
MDL Number: MFCD00044874
CAS Number: 12012-95-2
EINECS: 234-579-8

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR