Ammonium chloride ≥99.5%
Supplier: BeanTown Chemical
Synonyms:
Ammonium muriate, Salmiac
130480-2KG
130480-500G
BT130480-2KGEA
79
USD
BT130480-2KG
BT130480-500G
Ammonium chloride ≥99.5%
Ammonium chloride
CAS: 12125-02-9; EC No: 235-186-4; MDL No: MFCD00011420; RTECS: BP4550000
Granular; Linear Formula: NH4Cl; MW: 53.49
Melting Point: 340° (sublimes)
Hygroscopic
- RTECS: BP4550000
- Hygroscopic
- Granular
Formula:
NH₄Cl MW: 53.49 g/mol Melting Pt: 335 (subl.) |
MDL Number:
MFCD00011420 CAS Number: 12125-02-9 EINECS: 235-186-4 Merck Index: 13,00510 |
|
Specification Test Results
Assay | 99.5% |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...