2,5-Diamino-4,6-dichloropyrimidine ≥98%

Supplier: BeanTown Chemical

130465-1G 130465-5G
BT130465-1GEA 74 USD
BT130465-1G BT130465-5G
2,5-Diamino-4,6-dichloropyrimidine ≥98%
2,5-Diamino-4,6-dichloropyrimidine

CAS: 55583-59-0 Powder; Molecular Formula: C4H4Cl2N4; MW: 179.01


  • Powder

Formula: C₄H₄Cl₂N₄
MDL Number: MFCD00792652
CAS Number: 55583-59-0

Order Now

Specification Test Results

Assay 98%


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR