Diethyl propylmalonate ≥98%

Supplier: BeanTown Chemical

Synonyms: Diethyl 2-propylmalonate

130450-100G 130450-25G
BT130450-100GEA 130 USD
BT130450-100G BT130450-25G
Diethyl propylmalonate ≥98%
Diethyl propylmalonate

CAS: 2163-48-6; EC No: 218-492-2; MDL No: MFCD00009168 Liquid; Molecular Formula: C10H18O4 ; MW: 202.25 Boiling Point: 221-222°; Flash point: 92°C (198°F) Density (g/mL): 0.987 ; Refractive Index: 1.418


  • Refractive index: 1.418
  • Liquid

Formula: C₁₀H₁₈O₄
Boiling Pt: 221…222 °C
Density: 0.987 g/mL
Flash Pt: 92 °C (198 °F)
MDL Number: MFCD00009168
CAS Number: 2163-48-6
EINECS: 218-492-2

Order Now

Specification Test Results

Assay 99%


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR