(1-Ethoxycarbonylethylidene)triphenylphosphorane ≥97%

Supplier: BeanTown Chemical

130445-10G 130445-50G
BT130445-10GEA 79 USD
BT130445-10G BT130445-50G
(1-Ethoxycarbonylethylidene)triphenylphosphorane ≥97%
(1-Ethoxycarbonylethylidene)triphenylphosphorane

CAS: 5717-37-3; EC No: 227-215-4; MDL No: MFCD00009160 Crystalline/Powder; Molecular Formula: C23H23O2P; MW: 362.40 Melting Point: 158-162° Air Sensitive


  • Air Sensitive
  • Crystalline/Powder

Formula: C₂₃H₂₃O₂P
Melting Pt: 158…162 °C
MDL Number: MFCD00009160
CAS Number: 5717-37-3
EINECS: 227-215-4

Order Now

Specification Test Results

Assay 97%


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR