8-Bromoquinoline ≥98%

Supplier: BeanTown Chemical

130425-1G 130425-250MG
BT130425-1GEA 121 USD
BT130425-1G BT130425-250MG
8-Bromoquinoline ≥98%
8-Bromoquinoline

CAS: 16567-18-3; MDL No: MFCD00191859 Liquid; Molecular Formula: C9H6BrN ; MW: 208.05 Boiling Point: 112-113°/0.5 mmHg; Flash point: 110°C (230°F) Density (g/mL): 1.594; Refractive Index: 1.672 Light Sensitive


  • Refractive index: 1.672
  • Light Sensitive
  • Liquid

Formula: C₉H₆BrN
Boiling Pt: 112…113 °C/0.5 mmHg
Density: 1.594 g/mL
Flash Pt: >110 °C (>230 °F)
MDL Number: MFCD00191859
CAS Number: 16567-18-3

Order Now

Specification Test Results

Assay 98%


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR