Vitamin B12 (cyanocobalamin) ≥98%

Supplier: BeanTown Chemical

130400-1G 130400-25G 130400-5G
BT130400-1GEA 136 USD
BT130400-1G BT130400-25G BT130400-5G
Vitamin B12 (cyanocobalamin) ≥98%
Vitamin B12 (cyanocobalamin)

CAS: 68-19-9; EC No: 200-680-0; MDL No: MFCD00151092; RTECS: GG3750000 Crystalline/Powder; Molecular Formula: C63H88CoN14O14P; MW: 1355.38 Hygroscopic


  • RTECS: GG3750000
  • Hygroscopic
  • Crystalline/Powder

Formula: C₆₃H₈₈CoN₁₄O₁₄P
MW: 1355.38 g/mol
MDL Number: MFCD00151092
CAS Number: 68-19-9
EINECS: 200-680-0

Order Now

Specification Test Results

Assay 98%

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR