1H,1H,2H,2H-Tridecafluoro-1-octanol ≥97%
Supplier: BeanTown Chemical
Synonyms:
3,3,4,4,5,5,6,6,7,7,8,8,8-Tridecafluorooctan-1-ol, 1H,1H,2H,2H-Perfluoro-1-octanol, 3,3,4,4,5,5,6,6,7,7,8,8,8-Tridecafluoro-1-octanol, 1H,1H,2H,2H-Perfluorooctan-1-ol, 1H,1H,2H,2H-Perfluorooctanol
130395-10G
130395-250G
130395-50G
BT130395-10GEA
64.29
USD
BT130395-10G
BT130395-250G
BT130395-50G
1H,1H,2H,2H-Tridecafluoro-1-octanol ≥97%
1H,1H,2H,2H-Tridecafluoro-1-octanol
CAS: 647-42-7; EC No: 211-477-1; MDL No: MFCD00042143; RTECS: RH1460000
Liquid; Linear Formula: CF3(CF2)5(CH2)2OH; Molecular Formula: C8H5F13O; MW: 364.10
Boiling Point: 88-89°/28 mmHg; Flash point: 91°C (196°F)
Density (g/mL): 1.651; Refractive Index: 1.313
- RTECS: RH1460000
- Refractive index: 1.313
- Liquid
Formula:
C₈H₅F₁₃O MW: 364.11 g/mol Boiling Pt: 88…89 °C/28 mmHg Density: 1.651 g/mL Flash Pt: 91 °C (196 °F) |
MDL Number:
MFCD00042143 CAS Number: 647-42-7 EINECS: 211-477-1 |
Specification Test Results
Assay | 97% |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...