L(-)-Proline ≥99%

Supplier: BeanTown Chemical

Synonyms: (S)-(-)-ProlineH-Pro-OHL-Proline, H-Pro-OH

130390-100G 130390-25G 130390-500G
BT130390-100GEA 93 USD
BT130390-100G BT130390-25G BT130390-500G
L(-)-Proline ≥99%
L(-)-Proline

CAS: 147-85-3; EC No: 205-702-2; MDL No: MFCD00064318; RTECS: TW3584000 Powder; Molecular Formula: C5H9NO2; MW: 115.13 Melting Point: 228° (decomposes) Density (g/mL): 1.36 Hygroscopic


  • RTECS: TW3584000
  • Hygroscopic
  • Powder

Formula: C₅H₉NO₂
MW: 115.13 g/mol
Melting Pt: 228 °C (dec.)
Density: 1.36 g/mL
MDL Number: MFCD00064318
CAS Number: 147-85-3
EINECS: 205-702-2

Order Now

Specification Test Results

Assay 99%

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR