Calcium citrate tetrahydrate ≥96%
Supplier: BeanTown Chemical
130335-100G
130335-2.5KG
130335-500G
BT130335-100GEA
38
USD
BT130335-100G
BT130335-2.5KG
BT130335-500G
Calcium citrate tetrahydrate ≥96%
Calcium citrate tetrahydrate
CAS: 5785-44-4; EC No: 212-391-7; MDL No: MFCD00150786
Powder; Molecular Formula: C12H10Ca3O14·4H2O ; MW: 570.49
- Powder
Formula:
C₁₂H₁₀Ca₃O₁₄·4H₂O |
MDL Number:
MFCD00150786 CAS Number: 5785-44-4 EINECS: 212-391-7 |
Specification Test Results
Assay | 96% |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...