1H-Pyrazole-3-boronic acid pinacol ester ≥95%
Supplier: BeanTown Chemical
126525-1G
126525-5G
BT126525-1GEA
723
USD
BT126525-1G
BT126525-5G
1H-Pyrazole-3-boronic acid pinacol ester ≥95%
CAS: 844501-71-9; MDL No: MFCD07368044
Powder; Molecular Formula: C9H15BN2O2; MW: 194.04
- Powder
Formula:
C₉H₁₅BN₂O₂ |
MDL Number:
MFCD07368044 CAS Number: 844501-71-9 |
Specification Test Results
Assay | 95% |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...