EDTA tripotassium salt dihydrate ≥99%
Supplier: BeanTown Chemical
126505-250G
126505-50G
BT126505-250GEA
111
USD
BT126505-250G
BT126505-50G
EDTA tripotassium salt dihydrate ≥99%
EDTA tripotassium salt dihydrate
CAS: 65501-24-8; EC No: 241-543-5; MDL No: MFCD00149672; RTECS: MC0372000
Crystalline/Powder; Molecular Formula: C10H13K3N2O8·2H2O; MW: 442.55
Melting Point: 182° (decomposes)
- RTECS: MC0372000
- Crystalline/Powder
Formula:
C₁₀H₁₃K₃N₂O₈·2H₂O Melting Pt: 182 °C (dec.) |
MDL Number:
MFCD00149672 CAS Number: 65501-24-8 EINECS: 241-543-5 |
Specification Test Results
Assay | 99% |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...