Ethyl-trans-(ethoxymethylene)acetate ≥98%
Supplier: BeanTown Chemical
Synonyms:
Ethyl trans-3-ethoxyacrylate, Ethyl (E)-3-Ethoxyacrylate
126500-250G
126500-50G
BT126500-250GEA
305
USD
BT126500-250G
BT126500-50G
Ethyl-trans-(ethoxymethylene)acetate ≥98%
Ethyl-trans-(ethoxymethylene)acetate
CAS: 5941-55-9; MDL No: MFCD00009863
Liquid; Linear Formula: CH3CH2OCH=CHCOOCH2CH3; Molecular Formula: C7H12O3; MW: 144.17
Boiling Point: 195-196°; Flash point: 76°C (169°F)
Density (g/mL): 0.998; Refractive Index: 1.446
- Refractive index: 1.446
- Liquid
Formula:
C₇H₁₂O₃ Boiling Pt: 195…196 °C Density: 0.998 g/mL Flash Pt: 76 °C (169 °F) |
MDL Number:
MFCD00009863 CAS Number: 5941-55-9 |
Specification Test Results
Assay | 98% |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...