Tellurium ≥99.9999% (trace metals basis), broken ingot

Supplier: BeanTown Chemical

126490-100G 126490-25G 126490-500G
BT126490-100GEA 490 USD
BT126490-100G BT126490-25G BT126490-500G
Tellurium ≥99.9999% (trace metals basis), broken ingot
Tellurium

CAS: 13494-80-9; EC No: 236-813-4; MDL No: MFCD00134062; RTECS: WY2625000 Solid; Molecular Formula: Te; MW: 127.60 Melting Point: 450°; Boiling Point: 990° Density (g/mL): 6.24


  • RTECS: WY2625000
  • Solid

Formula: Te
MW: 127.6 g/mol
Boiling Pt: 990 °C
Melting Pt: 450 °C
Density: 6.24 g/mL
MDL Number: MFCD00134062
CAS Number: 13494-80-9
EINECS: 236-813-4
UN: 3284
ADR: 6.1,III

Order Now

Specification Test Results

Assay 99.9999%

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR