tert-Butyl (diethylphosphono)acetate ≥95%
Supplier: BeanTown Chemical
CAS: 27784-76-5; EC No: 000-000-0; MDL No: MFCD00075414
Liquid; Linear Formula: (CH3CH2O)P(O)CH2CO2C(CH3)3; Molecular Formula: C10H21O5P; MW: 252.25
Boiling Point: 100-103°/1.5 mmHg; Flash point:
- Refractive index: 1.431
- Moisture Sensitive
- Liquid
Formula:
C₁₀H₂₁O₅P Boiling Pt: 100…103 °C/1.5 mmHg Density: 1.074 g/mL Flash Pt: >110 °C (>230 °F) |
MDL Number:
MFCD00075414 CAS Number: 27784-76-5 |
Specification Test Results
Assay | 95% |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...