Polyethylene glycol 1,000
Supplier: BeanTown Chemical
Synonyms:
PEG 1,000, PEG 900-1100
126330-1KG
126330-250G
BT126330-1KGEA
81.8
USD
BT126330-1KG
BT126330-250G
Polyethylene glycol 1,000
Polyethylene glycol 1,000
CAS: 25322-68-3; MDL No: MFCD01779605; RTECS: TQ4025000
Powder; Linear Formula: H(OCH2CH2)nOH; MW ≈1,000
Melting Point: 35-40°; Flash point: 229°C (444°F)
Density (g/mL): 1.2
- RTECS: TQ4025000
- Powder
Formula:
H(OCH₂CH₂)nOH MW: 1000 g/mol Melting Pt: 35…40 °C Density: 1.2 g/mL Flash Pt: 229 °C (444 °F) |
MDL Number:
MFCD00081839 CAS Number: 25322-68-3 Merck Index: 13,07651 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...