Benzyl (R)-(-)-glycidyl ether ≥98%
Supplier: BeanTown Chemical
Synonyms:
(R)-2-((Benzyloxy)methyl)oxirane
126295-1G
126295-5G
BT126295-1GEA
76
USD
BT126295-1G
BT126295-5G
Benzyl (R)-(-)-glycidyl ether ≥98%
Benzyl (R)-(-)-glycidyl ether
CAS: 14618-80-5; MDL No: MFCD00068409; RTECS: TX2860030
Liquid; Molecular Formula: C10H12O2; MW: 164.20
Flash point: 110°C (230°F)
Density (g/mL): 1.077; Refractive Index: 1.517; Optical Rotation: [α]20/D -5.4°, c = 5 in toluene
- RTECS: TX2860030
- Optical rotation: [α]20/D -5.4°, c = 5 in toluene
- Refractive index: 1.517
- Liquid
Formula:
C₁₀H₁₂O₂ MW: 164.20 g/mol Density: 1.077 g/mL Flash Pt: 110 °C (230 °F) |
MDL Number:
MFCD00068409 CAS Number: 14618-80-5 |
Specification Test Results
Assay | 98% |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...