Dichloro(ethylenediamine)platinum(II)

Supplier: BeanTown Chemical

126285-1G 126285-250MG
BT126285-1GEA 263.97 USD
BT126285-1G BT126285-250MG
Dichloro(ethylenediamine)platinum(II)
Dichloro(ethylenediamine)platinum(II)

CAS: 14096-51-6; EC No: 237-943-4; MDL No: MFCD00000011 Crystalline/Powder; Linear Formula: PtCl2(H2NCH2CH2NH2); Molecular Formula: C2H8Cl2N2Pt; MW: 326.08 Hygroscopic


  • Hygroscopic
  • Crystalline/Powder

Formula: C₂H₈Cl₂N₂Pt
MW: 326.08 g/mol
MDL Number: MFCD00000011
CAS Number: 14096-51-6
EINECS: 237-943-4

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR