1,2,3,4-Tetramethylbenzene ≥95%
Supplier: BeanTown Chemical
126245-25G
126245-5G
BT126245-25GEA
432
USD
BT126245-25G
BT126245-5G
1,2,3,4-Tetramethylbenzene ≥95%
1,2,3,4-Tetramethylbenzene
CAS: 488-23-3; EC No: 207-673-1; MDL No: MFCD00008521; RTECS: DC0465000
Liquid; Molecular Formula: C10H14; MW: 134.22
Flash point: 68°C (154°F)
Density (g/mL): 0.9; Refractive Index: 1.519
- RTECS: DC0465000
- Refractive index: 1.519
- Liquid
Formula:
C₁₀H₁₄ Density: 0.9 g/mL Flash Pt: 68 °C (154 °F) |
MDL Number:
MFCD00008521 CAS Number: 488-23-3 EINECS: 207-673-1 |
Specification Test Results
Assay | 95% |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...