Potassium trifluoromethanesulfonate ≥98%
Supplier: BeanTown Chemical
126240-10G
126240-50G
BT126240-10GEA
56.78
USD
BT126240-10G
BT126240-50G
Potassium trifluoromethanesulfonate ≥98%
Potassium trifluoromethanesulphonate
CAS: 2926-27-4; MDL No: MFCD00042370
Crystalline; Linear Formula: CF3SO3K; MW: 188.17
Melting Point: 236°
Hygroscopic
- Hygroscopic
- Crystalline
Formula:
CF₃KO₃S MW: 188.17 g/mol Melting Pt: 236 °C |
MDL Number:
MFCD00042370 CAS Number: 2926-27-4 |
Specification Test Results
Assay | 98% |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...