Tetrabutylammonium hexafluorophosphate ≥98%

Supplier: BeanTown Chemical

Synonyms: Tetrabutylammonium hexafluorophosphate(V), Tetra-n-butylammonium hexafluorophosphate

126215-100G 126215-25G 126215-500G
BT126215-100GEA 227.29 USD
BT126215-100G BT126215-25G BT126215-500G
Tetrabutylammonium hexafluorophosphate ≥98%
Tetrabutylammonium hexafluorophosphate

CAS: 3109-63-5; EC No: 221-472-6; MDL No: MFCD00011748 Solid; Linear Formula: [CH3(CH2)3]4N(PF6); Molecular Formula: C16H236F6NP; MW: 387.44 Melting Point: 244-246° Moisture Sensitive


  • Moisture Sensitive
  • Solid

Formula: C₁₆H₃₆F₆NP
Melting Pt: 244…246 °C
MDL Number: MFCD00011748
CAS Number: 3109-63-5
EINECS: 221-472-6
UN: 3263
ADR: 8,III

Order Now

Specification Test Results

Assay 98%


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR