1-[(4-Ethoxyphenyl)ethynyl]-4-n-pentylbenzene ≥99%

Supplier: BeanTown Chemical

Synonyms: 1-Ethoxy-4-((4-pentylphenyl)ethynyl)benzene

126200-1G 126200-5G
BT126200-1GEA 123 USD
BT126200-1G BT126200-5G
1-[(4-Ethoxyphenyl)ethynyl]-4-n-pentylbenzene ≥99%
1-[(4-Ethoxyphenyl)ethynyl]-4-n-pentylbenzene

CAS: 95480-29-8; MDL No: MFCD01218020 Crystalline/Powder; Molecular Formula: C21H24O; MW: 292.42 Melting Point: 62°; Boiling Point: 178-180°/0.5 mmHg


  • Crystalline/Powder

Formula: C₂₁H₂₄O
MW: 292.422 g/mol
Boiling Pt: 178…180 °C/0.5 mmHg
Melting Pt: 62 °C
MDL Number: MFCD01218020
CAS Number: 95480-29-8

Order Now

Specification Test Results

Assay 99%

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR