Didodecyl sulphide ≥90%, Technical Grade

Supplier: BeanTown Chemical

Synonyms: Dodecyl sulphide

126150-100G 126150-25G 126150-5G
BT126150-100GEA 216 USD
BT126150-100G BT126150-25G BT126150-5G
Didodecyl sulphide ≥90%, Technical Grade
Didodecyl sulphide

CAS: 2469-45-6; EC No: 219-587-1; MDL No: MFCD00008975 Powder; Linear Formula: [CH3(CH2)11]2S ; Molecular Formula: C24H50S; MW: 370.73 Melting Point: 38-40°; Boiling Point: 260-263°/4 mmHg


  • Powder

Formula: C₂₄H₅₀S
MW: 370.73 g/mol
Boiling Pt: 465
Melting Pt: 40
MDL Number: MFCD00008975
CAS Number: 2469-45-6
EINECS: 219-587-1

Order Now

Specification Test Results

Assay 90%


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR