Methyl-1-aminocyclopropanecarboxylate hydrochloride

Supplier: BeanTown Chemical

126145-100MG 126145-500MG
BT126145-100MGEA 167 USD
BT126145-100MG BT126145-500MG
Methyl-1-aminocyclopropanecarboxylate hydrochloride
Methyl-1-aminocyclopropanecarboxylate hydrochloride

CAS: 72784-42-0; MDL No: MFCD00078879 Solid; Molecular Formula: C5H9NO2·HCl; MW: 151.59


  • Solid

Formula: C₅H₁₀ClNO₂
MDL Number: MFCD00078879
CAS Number: 72784-42-0

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR