Oleic acid ≥99%

Supplier: BeanTown Chemical

Synonyms: (9Z)-9-Octadecenoic acid

126125-100G 126125-25G 126125-5G
BT126125-100GEA 683.36 USD
BT126125-100G BT126125-25G BT126125-5G
Oleic acid ≥99%
Oleic acid

CAS: 112-80-1; EC No: 204-007-1; MDL No: MFCD00064242; RTECS: RG2275000 Liquid; Linear Formula: CH3(CH2)7CH=CH(CH2)7COOH; Molecular Formula: C18H34O2; MW: 282.47 Melting Point: 13-14°; Boiling Point: 194-195°/1.2 mmHg; Flash point: 110°C (230°F) Density (g/mL): 0.887; Refractive Index: 1.459 Air Sensitive


  • RTECS: RG2275000
  • Refractive index: 1.459
  • Air Sensitive
  • Liquid

Formula: C₁₈H₃₄O₂
MW: 282.47 g/mol
Boiling Pt: 194…195 °C/1.2 mmHg
Melting Pt: 13…14 °C
Density: 0.887 g/mL
Flash Pt: >110 °C (>230 °F)
MDL Number: MFCD00064242
CAS Number: 112-80-1
EINECS: 204-007-1
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=e5aecc5dce4d0318386a45d7adb7c5822631010a&vid=892a2df4ecab3679b06983b84cd27ad334ef961f&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Assay 99%

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR