Triammonium dodecamolybdophosphate hydrate

Supplier: BeanTown Chemical

126100-100G 126100-500G
BT126100-100GEA 62 USD
BT126100-100G BT126100-500G
Triammonium dodecamolybdophosphate hydrate
Triammonium dodecamolybdophosphate hydrate

CAS: 54723-94-3; EC No: 234-714-0; MDL No: MFCD00036312 Powder; Linear Formula: (NH4)3PMo12O40·xH2O; MW: 1876.35 (anhydrous)


  • Powder

Formula: H₁₂Mo₁₂N₃O₄₀P·H₂O
MDL Number: MFCD00150423
CAS Number: 54723-94-3
EINECS: 234-714-0

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR