4-Nitrobenzamide ≥98%
Supplier: BeanTown Chemical
126080-100G
126080-25G
BT126080-100GEA
176
USD
BT126080-100G
BT126080-25G
4-Nitrobenzamide ≥98%
4-Nitrobenzamide
CAS: 619-80-7; EC No: 210-613-7; MDL No: MFCD00007994; RTECS: CV5601000
Solid; Molecular Formula: C7H6N2O3 ; MW: 166.13
Melting Point: 199-201°
- RTECS: CV5601000
- Solid
Formula:
C₇H₆N₂O₃ MW: 166.14 g/mol Melting Pt: 199…201 °C |
MDL Number:
MFCD00007994 CAS Number: 619-80-7 EINECS: 210-613-7 |
Specification Test Results
Assay | 98% |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...