Molybdenum disilicide ≥99.5%
Supplier: BeanTown Chemical
Synonyms:
Molybdenum silicide
126020-250G
126020-50G
BT126020-250GEA
314
USD
BT126020-250G
BT126020-50G
Molybdenum disilicide ≥99.5%
Molybdenum disilicide
CAS: 12136-78-6; EC No: 235-231-8; MDL No: MFCD00151535
-325 Mesh Powder; Molecular Formula: MoSi2; MW: 152.11
Melting Point: 1870-2030°
Density (g/mL): 6.31
- -325 Mesh Powder
Formula:
MoSi₂ Melting Pt: 1870…2030 °C Density: 6.31 g/mL |
MDL Number:
MFCD00151535 CAS Number: 12136-78-6 EINECS: 235-231-8 |
Specification Test Results
Assay | 99.5% |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...