Manganese(II) sulfide ≥99.9%

Supplier: BeanTown Chemical

126000-100G 126000-25G 126000-5G
BT126000-100GEA 324 USD
BT126000-100G BT126000-25G BT126000-5G
Manganese(II) sulfide ≥99.9%
Manganese (II) sulphide

CAS: 18820-29-6; EC No: 242-599-3; MDL No: MFCD00014210 -325 Mesh Powder; Molecular Formula: MnS; MW: 87.00 Melting Point: 1610° (decomposes) Air Sensitive


  • Air Sensitive
  • -325 Mesh Powder

Formula: MnS
Melting Pt: 1610 °C (dec.)
MDL Number: MFCD00014210
CAS Number: 18820-29-6
EINECS: 242-599-3

Order Now

Specification Test Results

Assay 99.9%


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR