Cerium(IV) oxide ≥99.9%

Supplier: BeanTown Chemical

Synonyms: Ceric oxide, Cerium dioxide

125960-100G 125960-25G 125960-500G
BT125960-100GEA 235 USD
BT125960-100G BT125960-25G BT125960-500G
Cerium(IV) oxide ≥99.9%
Cerium(IV) oxide

CAS: 1306-38-3; EC No: 215-150-4; MDL No: MFCD00010933; RTECS: FK6310000 50-105nm APS Powder; Molecular Formula: CeO2; MW: 172.12 Density (g/mL): 7.13


  • RTECS: FK6310000
  • 50-105nm APS Powder

Formula: CeO₂
MW: 172.11 g/mol
Boiling Pt: 3468
Melting Pt: 2000
Density: 7.13 g/mL
MDL Number: MFCD00010933
CAS Number: 1306-38-3
EINECS: 215-150-4
Merck Index: 12,02035

Order Now

Specification Test Results

Assay 99.9%


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR