4-Amino-2,6-dichloropyridine ≥98%
Supplier: BeanTown Chemical
125910-1G
125910-250MG
125910-5G
BT125910-1GEA
41
USD
BT125910-1G
BT125910-250MG
BT125910-5G
4-Amino-2,6-dichloropyridine ≥98%
4-Amino-2,6-dichloropyridine
CAS: 2587-02-2; MDL No: MFCD00052832
Solid; Molecular Formula: C5H4Cl2N2; MW: 163.00
Melting Point: 169-173°
- Solid
Formula:
C₅H₄Cl₂N₂ Melting Pt: 169…173 °C |
MDL Number:
MFCD00052832 CAS Number: 2587-02-2 |
Specification Test Results
Assay | 98% |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...