4,4'-Dimethoxytrityl chloride ≥98%

Supplier: BeanTown Chemical

Synonyms: DMT-Cl, 4,4'-Dimethoxytriphenylmethyl chloride

125895-100G 125895-25G 125895-5G
BT125895-100GEA 521 USD
BT125895-100G BT125895-25G BT125895-5G
4,4'-Dimethoxytrityl chloride ≥98%
4,4'-Dimethoxytrityl chloride

CAS: 40615-36-9; EC No: 255-002-6; MDL No: MFCD00008409 Crystalline; Molecular Formula: C21H19ClO2; MW: 338.83 Melting Point: 119-123° Moisture Sensitive


  • Moisture Sensitive
  • Crystalline

Formula: C₂₁H₁₉ClO₂
MW: 338.8 g/mol
Melting Pt: 119…123 °C
MDL Number: MFCD00008409
CAS Number: 40615-36-9
EINECS: 255-002-6

Order Now

Specification Test Results

Assay 98%

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR