4-Bromo-3-fluoroanisole ≥97%

Supplier: BeanTown Chemical

125865-1G 125865-25G 125865-5G
BT125865-1GEA 49 USD
BT125865-1G BT125865-25G BT125865-5G
4-Bromo-3-fluoroanisole ≥97%
4-Bromo-3-fluoroanisole

CAS: 458-50-4; MDL No: MFCD01310983 Liquid; Molecular Formula: C7H6BrFO; MW: 205.03 Melting Point: 216-219°; Flash point: 85°C (185°F)


  • Liquid

Formula: C₇H₆BrFO
Melting Pt: 216…219 °C
Flash Pt: 85 °C (185 °F)
MDL Number: MFCD01310983
CAS Number: 458-50-4

Order Now

Specification Test Results

Assay 97%


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR