2-Fluoro-4-iodoaniline ≥99%

Supplier: BeanTown Chemical

125825-100G 125825-25G 125825-5G
BT125825-100GEA 389 USD
BT125825-100G BT125825-25G BT125825-5G
2-Fluoro-4-iodoaniline ≥99%
2-Fluoro-4-iodoaniline

CAS: 29632-74-4; MDL No: MFCD00011738 Solid; Molecular Formula: C6H5FIN ; MW: 237.01 Melting Point: 55-57°; Flash point: 110°C (230°F) Light Sensitive


  • Light Sensitive
  • Solid

Formula: C₆H₅FIN
MW: 237.02 g/mol
Melting Pt: 55…57 °C
Flash Pt: >110 °C (>230 °F)
MDL Number: MFCD00011738
CAS Number: 29632-74-4

Order Now

Specification Test Results

Assay 99%

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR