2-Amino-5-chlorobenzaldehyde ≥97%
Supplier: BeanTown Chemical
125815-1G
125815-5G
BT125815-1GEA
68
USD
BT125815-1G
BT125815-5G
2-Amino-5-chlorobenzaldehyde ≥97%
2-Amino-5-chlorobenzaldehyde
CAS: 20028-53-9; MDL No: MFCD02093936
Solid; Molecular Formula: C7H6ClNO; MW: 155.58
Air Sensitive
- Air Sensitive
- Solid
Formula:
C₇H₆ClNO MW: 155.58 g/mol |
MDL Number:
MFCD02093936 CAS Number: 20028-53-9 |
Specification Test Results
Assay | 97% |
Learn more
![](/m-us.vwr.com/en_US/images/about_vwr_275x315.jpg)
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...