6-Hydroxyhexanoic acid ≥95% may cont. variable amounts of dimer

Supplier: BeanTown Chemical

Synonyms: 6-Hydroxycaproic acid

125810-1G 125810-25G 125810-5G
BT125810-1GEA 75 USD
BT125810-1G BT125810-25G BT125810-5G
6-Hydroxyhexanoic acid ≥95% may cont. variable amounts of dimer
6-Hydroxyhexanoic acid

CAS: 1191-25-9; MDL No: MFCD00046560 Liquid; Molecular Formula: C6H12O3; MW: 132.16 Density (g/mL): 1.08; Refractive Index: 1.456


  • Refractive index: 1.456
  • Liquid

Formula: C₆H₁₂O₃
MW: 132.159 g/mol
Density: 1.08 g/mL
MDL Number: MFCD00046560
CAS Number: 1191-25-9

Order Now

Specification Test Results

Assay 95%


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR