Gibberellic acid ≥90%, Technical Grade
Supplier: BeanTown Chemical
Synonyms:
GA3, Gibberellin A3, (3S,3aS,4S,4aS,6S,8aR,8bR,11S)-6,11-Dihydroxy-3-methyl-12-methylene-2-oxo-4a,6-ethano-3,8b-prop-1-enoperhydroindeno[1,2-b]furan-4-carboxylic acid
125785-1G
125785-25G
125785-5G
BT125785-1GEA
52.13
USD
BT125785-1G
BT125785-25G
BT125785-5G
Gibberellic acid ≥90%, Technical Grade
Gibberellic acid
CAS: 77-06-5; EC No: 201-001-0; MDL No: MFCD00079329; RTECS: LY8990000
Powder; Molecular Formula: C19H22O6; MW: 346.37
Optical Rotation: [α]20/D +78±5°, c = 2% in methanol
- RTECS: LY8990000
- Optical rotation: [α]20/D +78±5°, c = 2% in methanol
- Powder
Formula:
C₁₉H₂₂O₆ MW: 346.38 g/mol Melting Pt: 234 |
MDL Number:
MFCD00079329 CAS Number: 77-06-5 EINECS: 201-001-0 Merck Index: 13,04430 |
Specification Test Results
Assay | 90% |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...