(S)-Di-tert-butyl-5-oxopyrrolidine-1,2-dicarboxylate ≥95%
Supplier: BeanTown Chemical
125770-1G
125770-250MG
BT125770-1GEA
403
USD
BT125770-1G
BT125770-250MG
(S)-Di-tert-butyl-5-oxopyrrolidine-1,2-dicarboxylate ≥95%
(S)-Di-tert-butyl-5-oxopyrrolidine-1,2-dicarboxylate
CAS: 91229-91-3; MDL No: MFCD03094770
Solid; Molecular Formula: C14H23NO5; MW: 285.34
- Solid
Formula:
C₁₄H₂₃NO₅ |
MDL Number:
MFCD03094770 CAS Number: 91229-91-3 |
Specification Test Results
Assay | 95% |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...