Ethyl phenylacetate ≥99%
Supplier: BeanTown Chemical
125745-100G
125745-2.5KG
125745-500G
BT125745-100GEA
27.27
USD
BT125745-100G
BT125745-2.5KG
BT125745-500G
Ethyl phenylacetate ≥99%
Ethyl phenylacetate
CAS: 101-97-3; MDL No: MFCD00009178; RTECS: AJ2824000
Liquid; Linear Formula: C6H5CH2CO2CH2CH3 ; Molecular Formula: C10H12O2; MW: 164.20
Boiling Point: 229°; Flash point: 98°C (208°F)
Density (g/mL): 1.03; Refractive Index: 1.497
- RTECS: AJ2824000
- Refractive index: 1.497
- Liquid
Formula:
C₁₀H₁₂O₂ MW: 164.2 g/mol Boiling Pt: 229 °C Density: 1.03 g/mL Flash Pt: 98 °C (208 °F) |
MDL Number:
MFCD00009178 CAS Number: 101-97-3 |
Specification Test Results
Assay | 99% |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...