2-Cyclopropyl-4-(4-fluorophenyl)-3-quinolinemethanol
Supplier: BeanTown Chemical
125705-1G
125705-5G
BT125705-1GEA
318
USD
BT125705-1G
BT125705-5G
2-Cyclopropyl-4-(4-fluorophenyl)-3-quinolinemethanol
2-Cyclopropyl-4-(4-fluorophenyl)-3-quinolinemethanol
CAS: 121660-11-5
Solid; Molecular Formula: C19H16FNO; MW: 293.33
Density (g/mL): 1.286
- Solid
Formula:
C₁₉H₁₆FNO Density: 1.286 g/mL |
MDL Number:
MFCD09032924 CAS Number: 121660-11-5 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...