Trimethylamine-N-oxide dihydrate ≥98%
Supplier: BeanTown Chemical
Synonyms:
Trimethylamine oxide dihydrate
125675-100G
125675-25G
125675-5G
BT125675-100GEA
121
USD
BT125675-100G
BT125675-25G
BT125675-5G
Trimethylamine-N-oxide dihydrate ≥98%
Trimethylamine N-oxide dihydrate
CAS: 62637-93-8; MDL No: MFCD00149077; RTECS: YH2850000
Crystalline; Linear Formula: (CH3)3NO·2H2O; MW: 111.14
Melting Point: 95-99°
- RTECS: YH2850000
- Crystalline
Formula:
C₃H₉NO·2H₂O MW: 111.14 g/mol (75.11 anhy) Melting Pt: 95…99 °C |
MDL Number:
MFCD00149077 CAS Number: 62637-93-8 |
Specification Test Results
Assay | 98% |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...