3-Cyclohexene-1-methanol ≥98%
Supplier: BeanTown Chemical
125615-100G
125615-25G
125615-5G
BT125615-100GEA
514
USD
BT125615-100G
BT125615-25G
BT125615-5G
3-Cyclohexene-1-methanol ≥98%
3-Cyclohexene-1-methanol
CAS: 1679-51-2; EC No: 216-847-6; MDL No: MFCD00001574
Liquid; Linear Formula: C6H9CH2OH; MW: 112.17
Boiling Point: 190-192°; Flash point: 93°C (163°F)
Density (g/mL): 0.961; Refractive Index: 1.484
- Refractive index: 1.484
- Liquid
Formula:
C₇H₁₂O Boiling Pt: 190…192 °C Density: 0.961 g/mL Flash Pt: 93 °C (163 °F) |
MDL Number:
MFCD00001574 CAS Number: 1679-51-2 EINECS: 216-847-6 |
Specification Test Results
Assay | 98% |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...