Potassium iodide ≥99.995% (trace metals basis)
Supplier: BeanTown Chemical
Synonyms:
Dipotassium diiodide, Kali iodide
125565-100G
125565-25G
125565-5G
BT125565-100GEA
434
USD
BT125565-100G
BT125565-25G
BT125565-5G
Potassium iodide ≥99.995% (trace metals basis)
Potassium iodide
CAS: 7681-11-0; EC No: 213-659-4; MDL No: MFCD00011405; RTECS: TT2975000
Powder; Linear Formula: KI; MW: 166.01
Melting Point: 681°; Boiling Point: 1330°
Density (g/mL): 3.13; Refractive Index: 1.677
Air Sensitive, Light Sensitive, Moisture Sensitive
- RTECS: TT2975000
- Refractive index: 1.677
- Air Sensitive, Light Sensitive, Moisture Sensitive
- Powder
Formula:
Kl MW: 166 g/mol Boiling Pt: 1330 °C Melting Pt: 681 °C Density: 3.13 g/mL |
MDL Number:
MFCD00011405 CAS Number: 7681-11-0 EINECS: 213-659-4 |
Specification Test Results
Assay | 99.995% |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...