Methyl-5-norbornene-2,3-dicarboxylic anhydride (mixture of isomers), Technical Grade
Supplier: BeanTown Chemical
Synonyms:
Methylnadic Anhydride, Methylnadic anhydride, Nadic Methyl Anhydride, Methyl-3,6-endomethylenetetrahydrophthalic Anhydride
125555-100G
125555-2.5KG
125555-500G
BT125555-100GEA
29
USD
BT125555-100G
BT125555-2.5KG
BT125555-500G
Methyl-5-norbornene-2,3-dicarboxylic anhydride (mixture of isomers), Technical Grade
Methyl-5-norbornene-2,3-dicarboxylic anhydride (mixture of isomers)
CAS: 25134-21-8; EC No: 246-644-8; MDL No: MFCD00167036
Liquid; Molecular Formula: C10H10O3; MW: 178.19
Boiling Point: 137-140°/10 mmHg; Flash point: 135°C (275°F)
Density (g/mL): 1.232; Refractive Index: 1.506
Moisture Sensitive
- Refractive index: 1.506
- Moisture Sensitive
- Liquid
Formula:
C₁₀H₁₀O₃ Boiling Pt: 137…140 °C/10 mmHg Density: 1.232 g/mL Flash Pt: 135 °C (275 °F) |
MDL Number:
MFCD00167036 CAS Number: 25134-21-8 EINECS: 246-644-8 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...