3-Fluoroisonicotinic acid ≥97%

Supplier: BeanTown Chemical

Synonyms: 3-Fluoropyridine-4-carboxylic acid

125530-1G 125530-250MG 125530-5G
BT125530-1GEA 58 USD
BT125530-1G BT125530-250MG BT125530-5G
3-Fluoroisonicotinic acid ≥97%
3-Fluoroisonicotinic acid

CAS: 393-53-3; MDL No: MFCD03789251 Solid; Molecular Formula: C6H4FNO2 ; MW: 141.10 Melting Point: 250° (decomposes)


  • Solid

Formula: C₆H₄FNO₂
MW: 141.1 g/mol
Melting Pt: 250 °C (dec.)
MDL Number: MFCD03789251
CAS Number: 393-53-3

Order Now

Specification Test Results

Assay 97%

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR