(+)-Dibenzoyl-D-tartaric acid monohydrate ≥99%

Supplier: BeanTown Chemical

Synonyms: (2S,3S)-2,3-Bis(benzoyloxy)succinic acid hydrate

125470-100G 125470-25G
BT125470-100GEA 149 USD
BT125470-100G BT125470-25G
(+)-Dibenzoyl-D-tartaric acid monohydrate ≥99%
(+)-Dibenzoyl-D-tartaric acid monohydrate

CAS: 80822-15-7; EC No: 241-097-1; MDL No: MFCD00150722 Powder; Molecular Formula: C18H14O8·H2O ; MW: 376.31 Melting Point: 150-153° Optical Rotation: [α]20/D +109°, c = 2% in ethanol Hygroscopic


  • Optical rotation: [α]20/D +109°, c = 2% in ethanol
  • Hygroscopic
  • Powder

Formula: C₁₈H₁₄O₈·1H₂O
MW: 376.33 g/mol (358.31 anhy)
Melting Pt: 150…153 °C
MDL Number: MFCD00150722
CAS Number: 80822-15-7
EINECS: 241-097-1

Order Now

Specification Test Results

Assay 99%

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR