Piperazine ferulate
Supplier: BeanTown Chemical
125455-1G
125455-5G
BT125455-1GEA
162
USD
BT125455-1G
BT125455-5G
Piperazine ferulate
Piperazine ferulate
CAS: 171876-65-6; MDL No:
MFCD03428534
Solid; Molecular Formula: C10H10O4·C4H10N2; MW: 280.32
- Solid
Formula:
C₁₄H₂₀N₂O₄ |
CAS Number:
171876-65-6 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...